siRNA Details
| Gene Symbol: LOC79999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291904.1 | NP_001278833.1 | 3 | 499 | ||
| XM_017025119.1 | XP_016880608.1 | 4 | 675 | |||
| Gene Symbol: GRAPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017024635.1 | XP_016880124.1 | 5 | 2598 | ||
| XM_017024636.1 | XP_016880125.1 | 4 | 2577 | |||
| GenBank mRNA | AK130829.1 | - | 11 | |||
| Gene Symbol: LOC102724624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524126.2 | XP_011522428.1 | 4 | 688 | ||
| Gene Symbol: LOC388436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291905.1 | NP_001278834.1 | 3 | 499 | ||
| GenBank mRNA | BC058012.1 | - | 3 | 511 | ||

LOC79999
Genomic Map