siRNA Details
| Gene Symbol: LOC100505797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134647.1 | 2, 3 | 260 | |||
| NR_134648.1 | 1, 2 | 86 | ||||
| GenBank mRNA | AW590150.1 | - | 1 | 195 | ||
| BG182684.1 | - | 1 | 14 | |||
| BX114234.1 | - | 2, 3 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505797| Gene Symbol: LOC100505797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134647.1 | 2, 3 | 260 | |||
| NR_134648.1 | 1, 2 | 86 | ||||
| GenBank mRNA | AW590150.1 | - | 1 | 195 | ||
| BG182684.1 | - | 1 | 14 | |||
| BX114234.1 | - | 2, 3 | 260 | |||