siRNA Details
| Gene Symbol: LOC100505797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134647.1 | 3 | 321 | |||
| NR_134648.1 | 2 | 147 | ||||
| GenBank mRNA | AW590150.1 | - | 1 | 134 | ||
| BX114234.1 | - | 3 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505797| Gene Symbol: LOC100505797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134647.1 | 3 | 321 | |||
| NR_134648.1 | 2 | 147 | ||||
| GenBank mRNA | AW590150.1 | - | 1 | 134 | ||
| BX114234.1 | - | 3 | 321 | |||