siRNA Details
| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145640.1 | NP_001139112.1 | 1 | 18 | ||
| NM_015174.1 | NP_055989.1 | 1 | 18 | |||
| NR_027149.1 | 1 | 18 | ||||
| XM_011527831.2 | XP_011526133.1 | 1 | 17 | |||
| GenBank mRNA | AK295037.1 | - | 1 | 17 | ||
| BC034005.1 | - | 1 | 22 | |||
| DA333602.1 | - | 1 | 18 | |||

ZFR2
Genomic Map