siRNA Details
| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007260.2 | NP_009191.1 | 7 | 469 | ||
| XM_005245728.4 | XP_005245785.1 | 7 | 941 | |||
| XM_017000165.1 | XP_016855654.1 | 7 | 836 | |||
| GenBank mRNA | AF090423.1 | - | 7 | 515 | ||
| AF098668.1 | - | 7 | 469 | |||
| AK024724.1 | - | 3 | 149 | |||
| BC017034.2 | - | 7 | 434 | 3845764 | ||
| BC017193.2 | - | 7 | 423 | 3138786 | ||
| Gene Symbol: LYPLA2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033250.1 | 1 | 458 | |||

LYPLA2
Genomic Map