siRNA Details
| Gene Symbol: LYPLA2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033250.1 | 1 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYPLA2P2| Gene Symbol: LYPLA2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033250.1 | 1 | 532 | |||