siRNA Details
| Gene Symbol: LOC101927151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI754970.1 | - | 5 | 517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927151| Gene Symbol: LOC101927151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI754970.1 | - | 5 | 517 | ||