siRNA Details
| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244847.1 | NP_001231776.1 | 3, 4 | 255 | ||
| NM_207392.2 | NP_997275.1 | 4, 5 | 297 | |||
| GenBank mRNA | AI184682.1 | - | 2, 3 | 200 | ||
| AJ293408.1 | - | 4, 5 | 258 | |||
| AY359007.1 | - | 4, 5 | 220 | |||
| BC130501.1 | - | 4, 5 | 297 | 40146532 | ||
| W95920.1 | - | 3, 4 | 164 | |||

KRTDAP
Genomic Map