siRNA Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004807.2 | NP_004798.3 | 2 | 1063 | ||
| GenBank mRNA | AB006179.1 | - | 2 | 1137 | ||
| AK293724.1 | - | 2 | 520 | |||
| AK295898.1 | - | 2 | 905 | |||
| BC001196.1 | - | 1 | 312 | |||
| BC096239.1 | - | 2 | 1039 | 40005544 | ||
| BC096240.1 | - | 2 | 1039 | 40005547 | ||
| BC099638.3 | - | 2 | 1039 | 40005546 | ||
| BC099639.1 | - | 2 | 1039 | 40005548 | ||

HS6ST1
Genomic Map