siRNA Details
| Gene Symbol: TRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013642.2 | NP_001013664.1 | 2 | 1167 | ||
| XM_005245867.3 | XP_005245924.1 | 2 | 1226 | |||
| GenBank mRNA | BC069216.1 | - | 2 | 1164 | 6376829 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRNP1| Gene Symbol: TRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013642.2 | NP_001013664.1 | 2 | 1167 | ||
| XM_005245867.3 | XP_005245924.1 | 2 | 1226 | |||
| GenBank mRNA | BC069216.1 | - | 2 | 1164 | 6376829 | |