siRNA Details
| Gene Symbol: LINC01398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121680.1 | 2 | 461 | |||
| GenBank mRNA | BC021746.1 | - | 2 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01398| Gene Symbol: LINC01398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121680.1 | 2 | 461 | |||
| GenBank mRNA | BC021746.1 | - | 2 | 465 | ||