siRNA Details
| Gene Symbol: LOC100996635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110621.1 | 1 | 153 | |||
| NR_110622.1 | 1 | 153 | ||||
| GenBank mRNA | HY120625.1 | - | 1 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996635| Gene Symbol: LOC100996635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110621.1 | 1 | 153 | |||
| NR_110622.1 | 1 | 153 | ||||
| GenBank mRNA | HY120625.1 | - | 1 | 135 | ||