siRNA Details
| Gene Symbol: FMO6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002601.1 | 5 | 1633 | |||
| GenBank mRNA | AK130511.1 | - | 5 | 1633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMO6P| Gene Symbol: FMO6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002601.1 | 5 | 1633 | |||
| GenBank mRNA | AK130511.1 | - | 5 | 1633 | ||