siRNA Details
| Gene Symbol: LOC102723886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135822.1 | 3 | 408 | |||
| GenBank mRNA | AW292250.1 | - | 2 | 292 | ||
| BE466517.1 | - | 2 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723886| Gene Symbol: LOC102723886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135822.1 | 3 | 408 | |||
| GenBank mRNA | AW292250.1 | - | 2 | 292 | ||
| BE466517.1 | - | 2 | 275 | |||