siRNA Details
| Gene Symbol: COA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012985.2 | NP_001013003.1 | 2 | 268 | ||
| NM_001206641.2 | NP_001193570.2 | 2 | 352 | |||
| NM_001301733.1 | NP_001288662.1 | 1 | 639 | |||
| GenBank mRNA | BC025793.1 | - | 2 | 175 | ||
| BC035925.1 | - | 2 | 265 | |||
| BC116455.1 | - | 2 | 268 | 4540930 | ||
| CB136291.1 | - | 2 | 321 | |||

COA6
Genomic Map