siRNA Details
| Gene Symbol: LOC388780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287682.1 | NP_001274611.1 | 1 | 366 | ||
| XM_011529236.2 | XP_011527538.1 | 1 | 857 | |||
| XM_017027847.1 | XP_016883336.1 | 1 | 857 | |||
| XM_017027848.1 | XP_016883337.1 | 1 | 857 | |||
| GenBank mRNA | AW006359.1 | - | 2 | 457 | ||

LOC388780
Genomic Map