siRNA Details
Gene Symbol: LINC00028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024358.1 | 2 | 365 | |||
GenBank mRNA | AI675392.1 | - | 3 | 219 | ||
BX280465.1 | - | 2 | 365 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024358.1 | 2 | 365 | |||
GenBank mRNA | AI675392.1 | - | 3 | 219 | ||
BX280465.1 | - | 2 | 365 |