siRNA Details
Gene Symbol: LINC00649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038883.2 | 1 | 536 | |||
NR_134559.1 | 1 | 536 | ||||
GenBank mRNA | AL532366.3 | - | 1 | 21 | ||
DA374118.1 | - | 1 | 446 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038883.2 | 1 | 536 | |||
NR_134559.1 | 1 | 536 | ||||
GenBank mRNA | AL532366.3 | - | 1 | 21 | ||
DA374118.1 | - | 1 | 446 |