siRNA Details
| Gene Symbol: MIATNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110543.1 | 2 | 277 | |||
| GenBank mRNA | AK026502.1 | - | 2 | 253 | ||
| BM709811.1 | - | 2 | 163 | |||
| BQ581362.1 | - | 2 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIATNB| Gene Symbol: MIATNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110543.1 | 2 | 277 | |||
| GenBank mRNA | AK026502.1 | - | 2 | 253 | ||
| BM709811.1 | - | 2 | 163 | |||
| BQ581362.1 | - | 2 | 293 | |||