siRNA Details
| Gene Symbol: TNRC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024843.1 | NP_001020014.1 | 2 | 206 | ||
| GenBank mRNA | BC028626.2 | - | 2 | 139 | 4826787 | |
| BE501815.1 | - | 4 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNRC6B| Gene Symbol: TNRC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024843.1 | NP_001020014.1 | 2 | 206 | ||
| GenBank mRNA | BC028626.2 | - | 2 | 139 | 4826787 | |
| BE501815.1 | - | 4 | 267 | |||