siRNA Details
| Gene Symbol: TRAPPC12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG683264.1 | - | 1 | 152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC12-AS1| Gene Symbol: TRAPPC12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG683264.1 | - | 1 | 152 | ||