siRNA Details
| Gene Symbol: MFSD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080473.1 | NP_001073942.1 | 12 | 1208 | ||
| XM_005264301.4 | XP_005264358.1 | 12 | 2269 | |||
| XM_017004063.1 | XP_016859552.1 | 11 | 2362 | |||
| XM_017004064.1 | XP_016859553.1 | 11 | 2347 | |||
| XM_017004065.1 | XP_016859554.1 | 10 | 2278 | |||
| XM_017004066.1 | XP_016859555.1 | 11 | 2330 | |||
| XM_017004067.1 | XP_016859556.1 | 10 | 2252 | |||
| XM_017004068.1 | XP_016859557.1 | 8 | 2087 | |||
| GenBank mRNA | BX647146.1 | - | 6 | 973 | ||

MFSD2B
Genomic Map