siRNA Details
Gene Symbol: TMEM247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145051.2 | NP_001138523.1 | 2 | 158 | ||
GenBank mRNA | AA431175.1 | - | 2 | 158 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145051.2 | NP_001138523.1 | 2 | 158 | ||
GenBank mRNA | AA431175.1 | - | 2 | 158 |