siRNA Details
| Gene Symbol: AAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014911.3 | NP_055726.3 | 22 | 13302 | ||
| GenBank mRNA | AK127187.1 | - | 1 | 161 | ||
| BC132937.1 | - | 1 | 147 | |||
| BC132939.1 | - | 1 | 147 | |||
| BX640963.1 | - | 1 | 575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AAK1| Gene Symbol: AAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014911.3 | NP_055726.3 | 22 | 13302 | ||
| GenBank mRNA | AK127187.1 | - | 1 | 161 | ||
| BC132937.1 | - | 1 | 147 | |||
| BC132939.1 | - | 1 | 147 | |||
| BX640963.1 | - | 1 | 575 | |||