siRNA Details
| Gene Symbol: BOLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035505.1 | NP_001030582.1 | 1, 2 | 101 | ||
| NM_212552.2 | NP_997717.2 | 1, 2 | 101 | |||
| GenBank mRNA | BC028282.1 | - | 1, 2 | 61 | ||
| BC042036.1 | - | 1, 2 | 97 | |||
| BU600037.1 | - | 106 | ||||
| DQ225187.1 | - | 1, 2 | 63 | |||

BOLA3
Genomic Map