siRNA Details
| Gene Symbol: SLC9A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011552.3 | NP_001011552.2 | 2 | 934 | ||
| XM_011511158.1 | XP_011509460.1 | 2 | 955 | |||
| GenBank mRNA | CR627411.1 | - | 2 | 935 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC9A4| Gene Symbol: SLC9A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011552.3 | NP_001011552.2 | 2 | 934 | ||
| XM_011511158.1 | XP_011509460.1 | 2 | 955 | |||
| GenBank mRNA | CR627411.1 | - | 2 | 935 | ||