siRNA Details
| Gene Symbol: LOC101928111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135575.1 | 2 | 241 | |||
| NR_135576.1 | 2 | 390 | ||||
| NR_135577.1 | 2 | 273 | ||||
| GenBank mRNA | AI653327.1 | - | 1 | 147 | ||
| BF434856.1 | - | 1 | 141 | |||
| BM704421.1 | - | 2 | 273 | |||

LOC101928111
Genomic Map