siRNA Details
| Gene Symbol: RBMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265060.2 | XP_005265117.1 | 1 | 108 | ||
| XM_005265061.2 | XP_005265118.1 | 1 | 108 | |||
| XM_005265062.1 | XP_005265119.1 | 1 | 108 | |||
| XM_017006178.1 | XP_016861667.1 | 1 | 108 | |||
| XM_017006179.1 | XP_016861668.1 | 1 | 108 | |||
| XM_017006180.1 | XP_016861669.1 | 1 | 108 | |||
| XM_017006181.1 | XP_016861670.1 | 1 | 108 | |||
| XM_017006182.1 | XP_016861671.1 | 1 | 108 | |||

RBMS3
Genomic Map