siRNA Details
| Gene Symbol: FAM212A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203370.1 | NP_976248.1 | 2 | 873 | ||
| XM_017006372.1 | XP_016861861.1 | 2 | 917 | |||
| GenBank mRNA | BC012170.1 | - | 2 | 873 | 4642270 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM212A| Gene Symbol: FAM212A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203370.1 | NP_976248.1 | 2 | 873 | ||
| XM_017006372.1 | XP_016861861.1 | 2 | 917 | |||
| GenBank mRNA | BC012170.1 | - | 2 | 873 | 4642270 | |