siRNA Details
| Gene Symbol: LINC01391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121649.1 | 2 | 180 | |||
| GenBank mRNA | AK128202.1 | - | 2 | 180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01391| Gene Symbol: LINC01391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121649.1 | 2 | 180 | |||
| GenBank mRNA | AK128202.1 | - | 2 | 180 | ||