siRNA Details
| Gene Symbol: ANKUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144960.2 | NP_001138432.1 | 3 | 750 | ||
| NM_001315505.1 | NP_001302434.1 | 3 | 722 | |||
| NM_001315506.1 | NP_001302435.1 | 3 | 750 | |||
| XM_011512798.1 | XP_011511100.1 | 3 | 247 | |||
| XM_011512799.1 | XP_011511101.1 | 3 | 481 | |||
| XM_011512802.1 | XP_011511104.1 | 3 | 481 | |||
| GenBank mRNA | AK098078.1 | - | 2 | 206 | ||
| AK304359.1 | - | 3 | 481 | |||
| BC171805.1 | - | 3 | 722 | |||

ANKUB1
Genomic Map