siRNA Details
| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110167.1 | 2 | 176 | |||
| NR_110168.1 | 2 | 129 | ||||
| GenBank mRNA | AW195433.1 | - | 2 | 336 | ||
| BF676681.1 | - | 2 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01213| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110167.1 | 2 | 176 | |||
| NR_110168.1 | 2 | 129 | ||||
| GenBank mRNA | AW195433.1 | - | 2 | 336 | ||
| BF676681.1 | - | 2 | 177 | |||