siRNA Details
| Gene Symbol: TMEM212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164436.1 | NP_001157908.1 | 1 | 21 | ||
| XM_011512817.1 | XP_011511119.1 | 1 | 21 | |||
| XM_017006376.1 | XP_016861865.1 | 1 | 31 | |||
| GenBank mRNA | DB199311.1 | - | 1 | 21 | ||

TMEM212
Genomic Map