siRNA Details
| Gene Symbol: LINC01063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132101.1 | 1 | 133 | |||
| GenBank mRNA | BU960446.1 | - | 1 | 136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01063| Gene Symbol: LINC01063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132101.1 | 1 | 133 | |||
| GenBank mRNA | BU960446.1 | - | 1 | 136 | ||