siRNA Details
Gene Symbol: LOC389199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203423.2 | NP_981968.1 | 1 | 738 | ||
GenBank mRNA | BC031673.1 | - | 1 | 738 | 5166037 |
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 1 | 185 | ||
NM_198595.2 | NP_940997.1 | 1 | 185 | |||
GenBank mRNA | AK290807.1 | - | 1 | 12 | ||
BI561136.1 | - | 1 | 183 | |||
DA605903.1 | - | 1 | 118 |
