siRNA Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145065.1 | NP_001138537.1 | 11 | 2619 | ||
| XM_011531936.1 | XP_011530238.1 | 12 | 2854 | |||
| XM_011531937.2 | XP_011530239.1 | 12 | 3255 | |||
| XM_011531938.2 | XP_011530240.1 | 12 | 2579 | |||
| XM_011531939.2 | XP_011530241.1 | 12 | 4079 | |||
| XM_017008194.1 | XP_016863683.1 | 12 | 2577 | |||
| GenBank mRNA | AB051467.1 | - | 11 | 2567 | ||

CCSER1
Genomic Map