siRNA Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146319.1 | NP_001139791.1 | 1 | 145 | ||
| XM_011543593.2 | XP_011541895.1 | 1 | 119 | |||
| GenBank mRNA | AK300154.1 | - | 1 | 90 | ||
| AK316288.1 | - | 1 | 109 | |||
| DC424558.1 | - | 1 | 145 | |||

GRAMD3
Genomic Map