siRNA Details
| Gene Symbol: SLC25A48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017009090.1 | XP_016864579.1 | 4 | 1060 | ||
| XM_017009092.1 | XP_016864581.1 | 4 | 1060 | |||
| Gene Symbol: LOC101930049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL700839.1 | - | 3 | 309 | ||

SLC25A48
Genomic Map