siRNA Details
Gene Symbol: LOC107986453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010130.1 | XP_016865619.1 | 2 | 2283 | ||
XM_017010131.1 | XP_016865620.1 | 2 | 2283 | |||
XM_017010132.1 | XP_016865621.1 | 2 | 2489 | |||
XM_017010133.1 | XP_016865622.1 | 2 | 2281 | |||
XM_017010134.1 | XP_016865623.1 | 2 | 2281 |
Gene Symbol: LOC101930049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW303377.1 | - | 2 | 426 |
