siRNA Details
| Gene Symbol: C5orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206966.2 | NP_996849.2 | 2 | 147 | ||
| XM_005268446.3 | XP_005268503.2 | 2 | 194 | |||
| XM_017009459.1 | XP_016864948.1 | 2 | 203 | |||
| GenBank mRNA | AK289403.1 | - | 2 | 147 | ||
| AY358656.1 | - | 2 | 111 | |||
| BC021680.1 | - | 2 | 151 | 3931172 | ||

C5orf46
Genomic Map