siRNA Details
| Gene Symbol: LOC105374972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134613.1 | 1 | 111 | |||
| NR_134614.1 | 1 | 111 | ||||
| GenBank mRNA | BM725887.1 | - | 1 | 57 | ||
| BX113657.1 | - | 1 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374972| Gene Symbol: LOC105374972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134613.1 | 1 | 111 | |||
| NR_134614.1 | 1 | 111 | ||||
| GenBank mRNA | BM725887.1 | - | 1 | 57 | ||
| BX113657.1 | - | 1 | 111 | |||