siRNA Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286550.1 | NP_001273479.1 | 2 | 170 | ||
| NM_207409.3 | NP_997292.2 | 2 | 170 | |||
| NR_104467.1 | 2 | 272 | ||||
| GenBank mRNA | AW665167.1 | - | 2 | 260 | ||
| AY358823.1 | - | 2 | 149 | |||
| CD513822.1 | - | 2 | 299 | |||

CLPSL2
Genomic Map