siRNA Details
Gene Symbol: GLYATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010904.1 | NP_001010904.1 | 6 | 896 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GLYATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010904.1 | NP_001010904.1 | 6 | 896 |