siRNA Details
| Gene Symbol: LOC100996634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010153.1 | XP_016865642.1 | 3 | 4336 | ||
| XM_017010154.1 | XP_016865643.1 | 3 | 4336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996634| Gene Symbol: LOC100996634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010153.1 | XP_016865642.1 | 3 | 4336 | ||
| XM_017010154.1 | XP_016865643.1 | 3 | 4336 | |||