siRNA Details
| Gene Symbol: LOC105378127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134591.1 | 3 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378127| Gene Symbol: LOC105378127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134591.1 | 3 | 330 | |||