siRNA Details
| Gene Symbol: LOC101929229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125432.1 | 2 | 112 | |||
| GenBank mRNA | AA448174.1 | - | 2 | 326 | ||
| AA995207.1 | - | 2 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929229| Gene Symbol: LOC101929229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125432.1 | 2 | 112 | |||
| GenBank mRNA | AA448174.1 | - | 2 | 326 | ||
| AA995207.1 | - | 2 | 324 | |||