siRNA Details
| Gene Symbol: SNTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287813.2 | NP_001274742.1 | 2 | 300 | ||
| NM_001321775.1 | NP_001308704.1 | 2 | 300 | |||
| GenBank mRNA | AK314084.1 | - | 2 | 237 | ||
| HY017401.1 | - | 2 | 301 | |||

SNTG1
Genomic Map