siRNA Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145160.1 | NP_001138632.1 | 3 | 400 | ||
| NM_003290.2 | NP_003281.1 | 2 | 415 | |||
| XM_005260042.3 | XP_005260099.1 | 2 | 197 | |||
| XM_006722865.2 | XP_006722928.1 | 3 | 400 | |||
| XM_011528231.2 | XP_011526533.1 | 3 | 313 | |||
| XM_011528232.1 | XP_011526534.1 | 2 | 1633 | |||
| XM_017027188.1 | XP_016882677.1 | 3 | 431 | |||
| XM_017027189.1 | XP_016882678.1 | 2 | 2266 | |||
| GenBank mRNA | AF186109.1 | - | 2 | 177 | ||
| AK023385.1 | - | 3 | 369 | |||
| AK300136.1 | - | 2 | 273 | |||
| AK301285.1 | - | 2 | 245 | |||
| AK315076.1 | - | 2 | 219 | |||
| BC037576.1 | - | 2 | 290 | 5582453 | ||
| BC067225.1 | - | 2 | 244 | 4772340 | ||
| BT019634.1 | - | 2 | 174 | |||
| DA032511.1 | - | 3 | 400 | |||
| FJ224305.1 | - | 2 | 224 | |||
| X05276.1 | - | 2 | 224 | |||

TPM4
Genomic Map