siRNA Details
| Gene Symbol: XKR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011720.1 | NP_001011720.1 | 5 | 1335 | ||
| NM_001287258.1 | NP_001274187.1 | 6 | 1453 | |||
| NM_001287259.1 | NP_001274188.1 | 6 | 1445 | |||
| NM_001287260.1 | NP_001274189.1 | 5 | 1189 | |||
| XM_006716447.3 | XP_006716510.1 | 3 | 685 | |||
| XM_011517524.2 | XP_011515826.1 | 5 | 1318 | |||
| XM_011517525.2 | XP_011515827.1 | 5 | 1180 | |||
| XM_017013404.1 | XP_016868893.1 | 4 | 1552 | |||
| GenBank mRNA | AY534247.1 | - | 5 | 1335 | ||
| BC137104.1 | - | 5 | 1253 | 9021101 | ||
| BC137105.1 | - | 5 | 1189 | 9021102 | ||
| BC151845.1 | - | 6 | 1412 | |||

XKR9
Genomic Map