siRNA Details
| Gene Symbol: C8orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243237.1 | NP_001230166.1 | 1 | 101 | ||
| XM_011517434.2 | XP_011515736.1 | 2 | 203 | |||
| XM_011517435.2 | XP_011515737.1 | 2 | 240 | |||
| GenBank mRNA | BC150551.1 | - | 1 | 101 | ||
| BC150552.1 | - | 1 | 102 | |||

C8orf89
Genomic Map